Learn More
AdipoGen Caspase-12 ms mAb 12G6
SDP
Supplier:  AdipoGen AG20T0141C100
Encompass_Preferred
Monoclonal Antibody. Recognizes an epitope within the p20 fragment of mouse caspase-12. Detects a band of ~51kDa by Western blot. Does not cross-react with mouse caspase-1 or -11. Isotype Rat IgG1kappa. Clone 12G6. Applications ELISA, FACS, WB. Liquid. In PBS containing 0.02% sodium azide. Caspase-12 is a member of the ICE/CED-3 family of caspases and is detected as a protein of ~51kDa. Overexpression of caspase-12 in cells induces apoptosis. Capase-12 is localized to the endoplasmatic reticulum ER and activated by ER stress and specifically cleaves and activates pro-caspase-9 and is also thought to be involved in the apoptosis induced by viral infection. Caspase-12-deficient mice are resistant to ER stress-induced apoptosis. Caspase-12 is probably expressed in many and perhaps all mouse tissues and is expressed at high levels in skeletal muscle, heart, lung and kidney and at moderate levels in brain and liver. In many humans caspase-12 is a pseudo gene and not expressed.
Catalog No. 50-256-3339
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.