Learn More
Description
Suggested positive control: HL-60. Predicted to react with Mouse and Rat.
Specifications
Specifications
| Antigen | Caspase 2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 18E809.3 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites, PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | CASP2 |
| Gene Accession No. | P42575 |
| Gene Alias | CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
