Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120941 Supplier Invitrogen™ Supplier No. PA5120941
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, Rat brain. Immunogen sequence: KLRLSTDTVE HSLDNKDGPV CLQVKPCTPE FYQTHFQLAY RLQSRPRGLA LVLSNVHFTG EKELEFRSGG DVDHSTLVTL FKLLGYDVHV LCDQTAQEMQ EKLQNFAQLP AHRVTDSCIV ALLSHGVEGA IYGVDGKLLQ LQEVFQLFDN ANCPSLQNKP KMFFIQACRG DETDRGVDQQ DGKNHAGSPG CEESDAGKEK LPKMRLPTRS DMICGYACLK GTAAMRNTKR GSWYIEALAQ VFSERACDMH VADMLVKVNA LIKDREGYAP GTEFHRCKEM SEYCSTLCRH LYLFPGHPPT.

Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CASP2
Gene Accession No. P29594, P42575, P55215
Gene Alias CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Symbols CASP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 153-452 of human Caspase-2 (NP_1167642).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12366, 64314, 835
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.