Learn More
MBL International Corporation APOPCYTO Caspase-3 Colorimetric Assay Kit
SDP
Supplier:  MBL International Corporation 4800
Encompass_Preferred
Size:100 Assays; APOPCYTO Caspase-3 Colorimetric Assay Kit detects caspase-3 activity in cell extract with DEVD-pNA as a substrate, which DEVD sequence is recognized by active caspase-3 selectively. APOPCYTO Caspase-3 Colorimetric Assay Kit is composed of reagents necessary for the detection, which are Cell Lysis buffer, Reaction buffer and so on. Caspase-3 activity can be detected easily and quickly using APOPCYTO Caspase-3 Colorimetric Assay Kit.Caspase 3 activity assay.
Catalog No. 50-446-13
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.