Learn More
Diagnostic Biosystems CASPASE 3 Caspase 3/CPP32
Supplier:  Diagnostic Biosystems RP09605
Encompass_Preferred
This antibody is specific to a 32 kDa protein, which is identified as Caspase 3. Caspase 3 is associated with induction of apoptosis. Caspase 3 is ubiquitously expressed and is synthesized as an inactive proenzyme.
Catalog No. 50-238-5489
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.