Learn More
Genetex Inc Caspase 3 Monoclonal Antibody [HMV307] HistoMAX™ (GTX639931) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639931
Encompass_Preferred
The Caspase 3 antibody [HMV307] HistoMAX™ (GTX639931) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human cervical adenocarcinoma tissue using GTX639931 Caspase 3 antibody [HMV307] HistoMAX. Adenocarcinoma with strong caspase 3 staining of tumor cells.
Catalog No. NC3669301