Learn More
Biocare Medical, LLC Caspase-3 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CP229C
Encompass_Preferred
Apoptosis has gained central importance in the study of many biological processes, including neoplasia, neurodegenerative diseases and development. The proteases that mediate apoptosis are called caspases (cysteinyl-aspartic acid proteases). Cleaved caspase-3 antibody detects endogenous levels of the large fragment of activated caspase-3, a protease that mediates apoptosis. Activation of caspase-3 requires proteolytic processing of its inactive zymogen into activated p17 and p12 subunits. Cleavage of caspase-3 requires aspartic acid at the P1 position. This antibody does not cross-react with other cleaved caspases.
Catalog No. 50-827-79
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.