Learn More
TRIBIOSCIENCE INC Caspase-3 Colorimetric Assay Kit, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2030
Encompass_Preferred
Caspase-3 Colorimetric Assay provides a simple and convenient approach for assaying the activity of caspases that recognize the sequence DEVD. The assay is based on spectrophotometric detection of the chromophore p-nitroaniline (pNA) after cleavage from the labeled substrate DEVD-pNA. The pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405-nm.
Catalog No. NC2886255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.