Learn More
Enzo Life Sciences Caspase-3 monoclonal antibody (31A1067) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804305C100
Encompass_Preferred
Clone: 31A1067. Isotype: IgG1. Immunogen: Recombinant human caspase-3. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes procaspase-3 and cleaved 17kDa catalytic subunit of caspase-3. Recognizes an epitope in the large (17kDa) domain subunit of caspase-3. Applications: IHC (FS), IHC (PS), WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS with 0.05% sodium azide.
Catalog No. 50-200-9860
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.