Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 3 p17 Monoclonal Antibody (31A1067)
GREENER_CHOICE

Catalog No. PIMA547699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547699 Supplier Invitrogen™ Supplier No. MA547699
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects pro Caspase-3 at 32 kDa* and the large subunit of the cleaved form of Caspase-3 at 14-21 kDa. The antibody can be used for western blot, immunohistochemistry and immuno-electron microscopy.

CASP3 (Caspase3 Apoptosis-Related Cysteine Protease), also known as YAMA, CPP32 or APOPAIN, is a caspase protein that interacts with caspase 8 and caspase 9. It is encoded by the CASP3 gene. The CASP3 protein is a member of the cysteine-aspartic acid protease (caspase) family. Tiso et al. (1996) used radiation hybrid mapping to localize the CPP32 gene to human chromosome 4q33-q35.1. Fernandes-Alnemri et al. (1994) found that overexpression of CPP32 in insect cells induced apoptosis. Coexpression of the 2 CPP32 subunits in insect cells also resulted in apoptosis. Tewari et al. (1995) showed that purified human ICE cleaved the Yama proenzyme into a proteolytically active form and that activated Yama cleaved PARP into the 85-kD apoptotic form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 3 p17
Applications Western Blot
Classification Monoclonal
Clone 31A1067
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CASP3
Gene Accession No. P42574, P55213, P70677
Gene Alias A830040C14Rik; AC-3; Apopain; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CC3; Cleaved Caspase 3; Cpp32; CPP-32; CPP32beta; cysteine protease CPP32; IRP; LICE; mldy; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein
Gene Symbols CASP3
Host Species Mouse
Immunogen Clone 31A1067 was developed against full-length recombinant human Caspase-3. The antibody recognizes an epitope in the large domain subunit (p17) of Caspase-3, and it will recognize pro Caspase-3 and the p17 cleavage fragment.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12367, 25402, 836
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.