Learn More
CPC Scientific Ac-Ile-Glu-Thr-Asp-AMC 5MG
SDP
Supplier:  CPC Scientific CASP056B
Encompass
ONE-LETTER SEQUENCE: Ac-IETD-AMCMOLECULAR FORMULA: C31H41N5O12MOLECULAR WEIGHT:675.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [219138-21-3], RESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry et al., J. Biol. Chem. , 272, 17907 (1997); Z. Han et al., J. Biol. Chem., 272, 13432 (1997)
Catalog No. 50-210-9019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.