Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Caspase-3 Substrate V, fluorogenic

For cellular and molecular biology applications

$164.70

Specifications

Color White to off-white
Form Lyophilized
Substrate Type Caspase Substrate
Molecular Weight (g/mol) 1360.38
Storage Requirements -30° to -10°C
View More Specs

Products 1
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
AAJ66767EXD
View Documents
Thermo Scientific Chemicals
J66767EXD
0.5mg
Each of 1 for $164.70
Order in Bulk

Request bulk or custom formats

Only null left
Add to Cart
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White to off-white
Caspase Substrate
-30° to -10°C
Lyophilized
1360.38
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.