Learn More
Invitrogen™ Caspase 6 Monoclonal Antibody (MCH2 14 1-190)

Description
A suggested positive control for this product is MCF-7 cell lysate.
Specifications
Specifications
| Antigen | Caspase 6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MCH2 14 1-190 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Casp6 |
| Gene Accession No. | P55212 |
| Gene Alias | apoptotic protease MCH-2; CASP6; CASP-6; caspase 6; caspase 6, apoptosis-related cysteine peptidase; caspase 6, apoptosis-related cysteine protease; caspase-6; Caspase-6 subunit p11; Caspase-6 subunit p18; mCASP-6; MCH2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.