Learn More
Invitrogen™ Caspase 7 Recombinant Rabbit Monoclonal Antibody (K01_1N93)

Description
Caspase 7 Recombinant Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Caspase 7 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1N93 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | CASP7 |
| Gene Accession No. | P97864 |
| Gene Alias | AI314680; Apoptotic protease Mch-3; Casp7; CASP-7; caspase 7; caspase 7, apoptosis-related cysteine peptidase; caspase 7, apoptosis-related cysteine protease; caspase-7; Caspase-7 subunit p11; Caspase-7 subunit p20; CMH-1; cysteine protease LICE2; ICE-IAP3; ICE-LAP3; ICE-like apoptotic protease 3; Lice2; mCASP-7; MCH3; OTTHUMP00000020511; OTTHUMP00000020513; OTTHUMP00000020514; RP11-211N11.6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.