Learn More
AdipoGen anti-Caspase-8 (human), mAb (C15) (50 µg)
SDP
Supplier:  AdipoGen AG20B0057C050
Encompass_Preferred
Monoclonal Antibody. Recognizes the p18 subunit of human caspase-8. Isotype: Mouse IgG2b. Clone: C15. Applications: ICC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Procaspase-8 belongs to the family of caspases. Binding of FasL to Fas leads to formation of a receptor complex at the cellular membrane, which was named DISC. The DISC consists of oligomerized receptors, the DD-containing adaptor molecule FADD, procaspase-8, procaspase-10 and c-FLIP. The DISC structure provides a platform for the oligomerization of procaspase-8 that allows two procaspase-8 homodimers to be in the close proximity leading to the initial activation of procaspase-8. At the first cleavage step, the N-terminal p43/p41 and the C-terminal p30 cleavage products are generated. At the second cleavage step, p43/p41 and p30 are processed to p10 and p18, respectively, which leads to the generation of the active caspase-8 heterotetramer (p18/p10)2.
Catalog No. 50-205-2968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.