Learn More
Invitrogen™ Caspase 9 (Cleaved Asp315) Polyclonal Antibody

Description
It is not recommended to aliquot this antibody. This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Specifications
Specifications
| Antigen | Caspase 9 (Cleaved Asp315) |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 6 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 50% glycerol, 100μg/mL BSA, 0.15M NaCl and no preservative; pH 7.5 |
| Gene | CASP9 |
| Gene Accession No. | P55211 |
| Gene Alias | APAF3; APAF-3; apoptosis-related cysteine protease; apoptotic protease activating factor 3; apoptotic protease MCH-6; Apoptotic protease-activating factor 3; cas9; CASP9; CASP-9; caspase 9; caspase 9, apoptosis-related cysteine peptidase; caspase-9; Caspase-9 subunit p10; Caspase-9 subunit p35; ICE-LAP6; ICE-like apoptotic protease 6; MCH6; OTTHUMP00000002324; PPP1R56; protein phosphatase 1, regulatory subunit 56 |
| Gene Symbols | CASP9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.