Learn More
Invitrogen™ Caspase 9 Polyclonal Antibody

Description
Recommended positive controls: HeLa (30 μM cisplatin treatment for 24 hr), NTUB1. Predicted reactivity: Dog (80%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Caspase 9 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.55 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | CASP9 |
| Gene Accession No. | P55211 |
| Gene Alias | 25 kDa caspase-9 dominant negative protein; AI115399; APAF3; APAF-3; apoptotic protease activating factor 3; apoptotic protease MCH-6; Apoptotic protease-activating factor 3; AW493809; cas9; CASP 9; CASP9; CASP-9; Casp9_v1; Casp-9-CTD; caspase 9; caspase 9, apoptosis-related cysteine peptidase; caspase 9, apoptosis-related cysteine protease; Caspase9; caspase-9; caspase-9 dominant negative form; Caspase-9 precursor; Caspase-9 subunit p10; Caspase-9 subunit p35; CASPASE-9c; caspase-9-carboxyl-terminal divergent; CTD; Ice-Lap6; ICE-like apoptotic protease 6; Mch6; OTTHUMP00000002324; PPP1R56; protein phosphatase 1, regulatory subunit 56; RNCASP9; RP11-265F14.3 |
| Gene Symbols | CASP9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.