Learn More
Enzo Life Sciences Caspase-inhibitor set III (1 Set)
Supplier: Enzo Life Sciences ALX850227KI01
Synthetic peptides that irreversibly inhibit the activity of each of the caspase-family proteases. YVAD is the recognition sequence for caspase-1/ICE, VDVAD is the recognition sequence for caspase-2, DEVD is the recognition sequence for caspase-3/CPP32, WEHD is the recognition sequence for caspase-5, VEID is the recognition sequence for caspase-6/Mch2, IETD is the recognition sequence for caspase-8/FLICE, LEHD is the recognition sequence for caspase-9, and VAD is the recognition sequence for a caspase(s) that functions upstream of most other caspases. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.