Learn More
Nasco CasPeR The CPR Dog
SDPSCI_ED
Catalog No. S01258 Shop All Simulaids, Inc. Products
Click to view available options
For Use With (Application):
CPR Training
Catalog No. For Use With (Application)
S01258 CPR Training
1 options
Catalog No. S01258 Supplier Nasco Supplier No. 1005000
CasPeR The CPR Dog serves both veterinarian and lay persons in the instruction of CPR.
  • CasPeR The CPR Dog can demonstrate the correct:
    • Rescue position for administering CPR
    • Tongue movement for clearing the airway and sealing the oral space
    • Position for checking femoral pulse
    • Hand position for compressions
    • Procedure and depth of compressions
    • Procedure for rescue breathing via mouth-to-snout
  • The individual-use airway/lung system is a proven method for eliminating cross-contamination between students and each student discards the airway after the day's instruction ends.
  • The system needs no tools to install and removes with a gentle tug. When the student ventilates the manikin, the dog's chest will rise in proportion to the volume of air instilled.
  • CasPeR comes in a box with a convenient carry handle, is supplied with 100 airway systems, and has a squeeze bulb for generating femoral pulses
Specifications
Product Type Basic Sanitary CPR Dog
For Use With (Application) CPR Training
WARNING: Cancer - www.P65Warnings.ca.gov