Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
Catalog No. B851739GY Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851739GY Gray
B851739BE Beige
B851739BL Blue
B851739GN Green
B851739LA Lavender
B851739LG Light Green
B851739LI Lilac
B851739OR Orange
B851739PK Pink
B851739TN Tan
B851739WH White
B851739YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. B851739GY Supplier Epredia™ Supplier No. B851739GY
Add to Cart
Add to Cart
For routine tissue samples, these loose-packed cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Pore Style Slots
Closure Included Yes
Quantity 1000/Cs.
Closure Type Separate Lid, Front Hinge
Color Gray
Product Type Slotted Cassette