Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Cast Material Stacking Box
SDP

Catalog No. 21377548
Change view
Click to view available options
Width (English):
4 in.
5 in.
6 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
21-377-548 4 in.
21-377-549 5 in.
21-377-550 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-548 Supplier Poltex Supplier No. CASTBXSTK4
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

Stackable box for holding cast material with protrusions on top that fit into pockets on base for secure stackablity.

  • Stackable box for cast material
  • Durable and cleanable PETG construction
  • Protrusions on top fit into pockets on base for secure stackablity
  • Lid lifts off for full access to cast material
  • Choose from: 4, 5, or 6 in. wide

Specifications

Type Dispensing Bin
Material PETG
Color Clear
Shape Rectangular
Autoclavable Not Autoclavable
Width (English) 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.