Learn More
Labconco™ Caster Kits
SDP
Catalog No. 16101153 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Base Kit
Conversion Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-101-153 Base Kit
10-269-356 Conversion Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-101-153 Supplier Labconco™ Supplier No. 3784000
Add to Cart
Add to Cart
Replaces fixed feet included with electric and manual hydraulic lift base stands to add mobility
  • Includes four 4.5 in. dia. (12.7cm) nonmarking polyurethane casters with toe locks and bearings for quiet movement
  • Load rating: 250 lb. (113.4kg) per caster
  • Installation required
Specifications
For Use With (Equipment) Adjustable Base Stand
Includes Four 4.5 in. diameter Wheels with toe locks (113.4kg [250 lb.] load rating per caster)
Product Type Base Kit
WARNING: Cancer - www.P65Warnings.ca.gov