Learn More
TARGETMOL CHEMICALS INC CASTICIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S1373
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Casticin (Vitexicarpin) can significantly reduce vascular inflammation through inhibition of ROS-NF-(kappa)B pathway in vascular endothelial cells. 2. Casticin may become a potential leading drug in the therapy of prostate carcinoma. purity: 99%
Catalog No. 50-257-6379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.