Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Castor Oil, USP, Spectrum™ Chemical
SDP

Catalog No. 18601511 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-511 20 L Amber Glass Bottle
18-601-509 500 mL Amber Glass Bottle
18-601-510 4 L Amber Glass Bottle
18-601-512 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-511 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA20820LTMT
Add to Cart
Add to Cart

CA208, 8001-79-4

Spectrum™ Chemical Castor Oil, USP is used as a stimulant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
InChI Key ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
IUPAC Name 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC

Specifications

CAS Max % 100%
Boiling Point 313°C
Packaging Amber Glass Bottle
Quantity 20 L
Grade USP
Physical Form Liquid
Chemical Name or Material Castor Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.