Learn More
Surgical Design™ Castroviejo Scissors and Needle Holders - TC, Straight and Curved
Catalog No. 21124696 Shop All Surgical Design Products
Change view
Click to view available options
Tip Style:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
21-124-696 Curved
21-124-695 Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-696 Supplier Surgical Design™ Supplier No. CSS5504TC
Add to Cart
Add to Cart
Castroviejo scissors and needle holders with TC inserts provide maximum control for ophthalmic, micro, vascular, and fine dissection work.
  • Four instrument options available: Castroviejo Scissors TC Straight, Castroviejo Scissors TC Curved, Castroviejo Needle Holder TC Straight, Castroviejo Needle Holder, TC Curved
  • Tungsten carbide (TC) inserts provide exceptional durability and improved grip or cutting efficiency
  • Fine, micro-precision tips designed for delicate surgical and autopsy applications
  • Smooth spring-action handle allows controlled, low-fatigue operation
  • Ideal for ophthalmic, vascular, microsurgical, dermatologic, and forensic/postmortem use
  • Crafted from surgical-grade stainless steel with excellent corrosion resistance
  • Lightweight and easy to maneuver for precise control
  • Fully autoclavable and reusable
Specifications
Product Type Needle Holder Driver
Autoclavable Autoclavable
Coating Tungsten Carbide
Color Silver
Material Stainless Steel
Latex Free Yes
Tip Style Curved
Length (English) 5.5 in.
Length (Metric) 14 cm
Sterility Non-sterile