Learn More
Abcam Catalase Activity Assay Kit, Immunocapture, 96 Tests
Description
- Assay Type: Enzyme activity
- Assay Time: 6 hr
- Range: 1 μg/mL to 1,000 μg/mL
- Sample Types: Cell culture extracts, Tissue extracts
- Applications: Enzyme activity assay
- Reactive Species: Human, Mouse, Rat
- Citations: 12 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | 96-well Plate Kit |
| Detection Method | Colorimetric |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Target Molecule | Catalase |
| No. of Reactions | 96 Tests |
| Cross Reactivity | Human, Mouse, Rat |
| Sample Type | Cell Culture Extracts, Tissue extracts |
| Quantity | 96 Tests |
| For Use With (Application) | Enzyme activity assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.