Learn More
Cayman Chemical Catalase Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 70700296WELLS
Encompass_Preferred
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilizes the peroxidatic function of catalase for determination of enzyme activity. The assay can be used to measure catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.
Catalog No. NC9361871
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.