Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Catalase (Bovine)

Catalog No. 89938126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-938-126 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-938-126 Supplier Abnova™ Supplier No. P5218
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number P00432
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 531682
Molecular Weight (g/mol) 240kDa
Name Catalase (Bovine)
Preparation Method Native protein purified from Bovine Liver
Quantity 2 g
Storage Requirements Store at -20°C on dry atmosphere.
Regulatory Status RUO
Gene Alias MGC128112
Common Name CAT
Gene Symbol CAT
Assays Catalase Assay; Free radical research; Deoderization; Decomposition of residual hydrogen peroxide after bleaching woven and knitted cotton fabrics before drying; Cysteamine determination; Gluconic and glycolic acid production; Cleaning silicon and semiconductor plates.
Biological Activity ≥3,000 units per mg protein. One Unit decomposes one micromole of hydrogen peroxide per minute at 25°C, pH 7.0.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Liver
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.