Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase from Bovine Liver

Catalog No. AAJ1288503 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
10 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ1288503 1 g
AAJ12885ME 500 mg
AAJ1288506 5 g
AAJ1288509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ1288503 Supplier Thermo Scientific Supplier No. J1288503
  • $120.50
    $102.65 / Each of 1

    Save $17.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Derived from Bovine liver

  • One unit of enzyme decomposes 1μmol H2O2/min at 25°C at pH 7.0
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥3,000 U/mg
pH 7
Content And Storage -20°C, Protect from moisture
Source Bovine Liver
Quantity 1 g
Product Type Catalase
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.