Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase from Bovine Liver

Catalog No. AAJ12885ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
10 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ12885ME 500 mg
AAJ1288503 1 g
AAJ1288506 5 g
AAJ1288509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ12885ME Supplier Thermo Scientific Supplier No. J12885ME
Add to Cart

Derived from Bovine liver

  • One unit of enzyme decomposes 1μmol H2O2/min at 25°C at pH 7.0
TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥3,000 U/mg
pH 7
Content And Storage -20°C, Protect from moisture
Source Bovine Liver
Quantity 500 mg
Product Type Catalase
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.