Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase from Human Erythrocytes (E.C.1.11.1.6.)

Catalog No. MP219134183 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 kU
25 kU
250 kU
50 kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219134183 250 kU
MP219134150 50 kU
MP219134180 100 kU
ICN19134125 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219134183 Supplier MP Biomedicals™ Supplier No. 0219134183
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Negative for HBsAg and HIV antibodies

Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.

Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).

Activity: approx. 160,000U/mg protein. Supplied as frozen solution of 1mg/mL in 1.5mM sodium phosphate, 20mM NaCl, pH 7.0 composed of four identical subunits. Unit definition: One unit is defined as the amount of enzyme that will decompose 1μmole of H2O2 per minute at 25°C, pH 7.0.

Ref.: Bonaventura, J., et al., Arch. Biochem. Biophysics, 150: 606 (1972).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Protecting cells against oxidative damage to proteins, lipids and nucleic acids
Storage Buffer Frozen solution of 1mg/mL in 1.5mM Sodium Phosphate, 20mM NaCl, pH 7.0
Content And Storage -20°C
Source Human erythrocyte
Quantity 250 kU
Product Type Catalase
Molecular Weight (g/mol) 256000
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.