Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase, MP Biomedicals™

Catalog No. MP219031104 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219031104 4 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219031104 Supplier MP Biomedicals™ Supplier No. 0219031104
Add to Cart
Add to Cart

Catalase is used for the removal of peroxides, the generation of oxygen.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Content And Storage -20°C, Desiccate
Source Aspergillus niger
Quantity 4 mU
Molecular Weight (g/mol) ∽240,000
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.