Learn More
Biomatik Corporation CYP4F11 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07575
Encompass_Preferred
The CYP4F11 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Phylloquinone omega-hydroxylase CYP4F11 protein (38-524AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:1000-1:5000, IF:1:50-1:200. Alternative Names: Phylloquinone omega-hydroxylase CYP4F11, 3-hydroxy fatty acids omega-hydroxylase CYP4F11, Cytochrome P450 4F11, CYPIVF11, CYP4F11. UniProt: Q9HBI6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.