Learn More
Biomatik Corporation NPY6R anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07932
Encompass_Preferred
Npy6R Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NPY6R. The Npy6R Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Putative Neuropeptide Y Receptor Type 6 Protein (1-39Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: NPY6R. Accession Number: Q99463. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.