Learn More
Biomatik Corporation FKBP6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08441
Encompass_Preferred
The FKBP6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Inactive peptidyl-prolyl cis-trans isomerase FKBP6 protein (101-327AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Inactive peptidyl-prolyl cis-trans isomerase FKBP6, Inactive PPIase FKBP6, 36 kDa FK506-binding protein, 36 kDa FKBP, FKBP-36, FK506-binding protein 6, FKBP-6, Immunophilin FKBP36, FKBP6, FKBP36. UniProt: O75344. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8024
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.