Learn More
Biomatik Corporation GPR17 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09287
Encompass_Preferred
The GPR17 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Uracil nucleotide/cysteinyl leukotriene receptor protein (1-36AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Uracil nucleotide/cysteinyl leukotriene receptor, UDP/CysLT receptor, GPR17, G-protein coupled receptor 17, P2Y-like receptor, R12. UniProt: Q13304. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-8870
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.