Learn More
Biomatik Corporation Glutaminyl Peptide Cyclotransferase (QPCT) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU22013
Encompass_Preferred
The Glutaminyl Peptide Cyclotransferase (QPCT) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: QPCT (Val29-Leu361). Applications: WB, IHC. Alternative Names: GCT; QC; EC; Glutaminyl Cyclase; Glutaminyl-tRNA cyclotransferase. UniProt: Q16769. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9779
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.