Learn More
Biomatik Corporation Alpha-2-Macroglobulin (a2M) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26678
Encompass_Preferred
The Alpha-2-Macroglobulin (a2M) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: a2M (Ser616-Val856). Applications: WB, IHC. Alternative Names: A2MG; A2mg; CPAMD5; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5. UniProt: P01023. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9926
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.