Learn More
Biomatik Corporation Platelet/Endothelial Cell Adhesion Molecule (PECAM1) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28051
Encompass_Preferred
Platelet/Endothelial Cell Adhesion Molecule (Pecam1) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Platelet/Endothelial Cell Adhesion Molecule (PECAM1). The Platelet/Endothelial Cell Adhesion Molecule (Pecam1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Arg459~Glu624. Applications: WB, IHC, ICC, IP. Target Synonyms: Platelet/Endothelial Cell Adhesion Molecule (PECAM1). Accession Number: Q3SWT0. Gene ID: 29583. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0029
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.