Learn More
Biomatik Corporation Tumor Necrosis Factor Alpha (TNFa) anti-Chicken, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28503
Encompass_Preferred
Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Tumor Necrosis Factor Alpha (TNFa). The Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Arg1~Phe73. Applications: WB, IHC, ICC, IP. Target Synonyms: Tumor Necrosis Factor Alpha (TNFa). Accession Number: AEW10516.1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.