Learn More
Biomatik Corporation Cyclin D1 (CCND1) anti-Chicken, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29276
Encompass_Preferred
The Cyclin D1 (CCND1) Monoclonal Antibody is raised in Mouse. Species Reactivity: Chicken. Immunogen: Met1-Ile292. Applications: WB. Alternative Names: CCN-D1; BCL1; PRAD1; U21B31; G1/S-Specific Cyclin-D1; Parathyroid Adenomatosis 1; B-Cell CLL/Lymphoma 1; G1/S-Specific Cyclin D1; PRAD1 oncogene. UniProt: P55169. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.