Learn More
Biomatik Corporation Heat Shock 60kD Protein 1, Chaperonin (HSPD1) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29304
Encompass_Preferred
The Heat Shock 60kD Protein 1, Chaperonin (HSPD1) Monoclonal Antibody is raised in Mouse. Species Reactivity: Human. Applications: WB, IHC, ICC, IP. Alternative Names: HSP60; GROEL; CPN60; HuCHA60; Heat Shock Protein 60; Spastic Paraplegia 13,Autosomal Dominant; 60 kDa chaperonin; Mitochondrial matrix protein P1; P60 lymphocyte protein. UniProt: P10809. Concentration: 1 mg/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0170