Learn More
Biomatik Corporation MYH9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07304
Encompass_Preferred
Myh9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MYH9. The Myh9 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Myosin-9 Protein (2-241Aa). Applications: ELISA, IHC, IF. Target Synonyms: MYH9. Accession Number: P35579. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6890