Learn More
Biomatik Corporation PKM anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07314
Pkm Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PKM. The Pkm Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Pyruvate Kinase Pkm Protein (2-192Aa). Applications: ELISA, IHC, IF. Target Synonyms: PKM. Accession Number: P14618. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.