Learn More
Biomatik Corporation R-Spondin 3 (Rspo3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21332
Encompass_Preferred
R-Spondin 3 (Rspo3) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to R-Spondin 3 (RSPO3). The R-Spondin 3 (Rspo3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln22~His272. Applications: WB, IHC, ICC, IP. Target Synonyms: R-Spondin 3 (RSPO3). Accession Number: Q9BXY4. Gene ID: 84870. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.