Learn More
Biomatik Corporation Cytochrome P450 17A1 (Cyp17A1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24838
Encompass_Preferred
Cytochrome P450 17A1 (Cyp17A1) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Cytochrome P450 17A1 (CYP17A1). The Cytochrome P450 17A1 (Cyp17A1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Thr201~Thr507. Applications: WB, IHC, ICC, IP. Target Synonyms: Cytochrome P450 17A1 (CYP17A1). Accession Number: P11715. Gene ID: 25146. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9363