Learn More
Biomatik Corporation Interleukin 35 (Il35) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24863
Encompass_Preferred
Interleukin 35 (Il35) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interleukin 35 (IL35). The Interleukin 35 (Il35) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: The IL35 complex composed of IL27B subunit (Arg21~Lys229) and the mature form of human IL12A (Arg23~Ser219) linked by a poly peptide linker (GGGGS)3. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 35 (IL35). Accession Number: P29459. Gene ID: 3592. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9367
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.