Learn More
Biomatik Corporation Sialic Acid Binding Ig Like Lectin 14 (Siglec14) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21055
Encompass_Preferred
Sialic Acid Binding Ig Like Lectin 14 (Siglec14) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sialic Acid Binding Ig Like Lectin 14 (SIGLEC14). The Sialic Acid Binding Ig Like Lectin 14 (Siglec14) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln24~Leu137. Applications: WB, IHC. Target Synonyms: Sialic Acid Binding Ig Like Lectin 14 (SIGLEC14). Accession Number: Q08ET2. Gene ID: 100049587. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9186
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.