Learn More
Biomatik Corporation Recombinant Hyaluronan Binding Protein 1 (HABP1), 100UG
Supplier:  Biomatik Corporation RPU41945
Encompass_Preferred
Recombinant Human Complement component 1 Q subcomponent-binding protein, mitochondrial (C1QBP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Complement component 1 Q subcomponent-binding protein, mitochondrial (C1QBP). Target Synonyms: HABP1; GC1QBP; SF2p32; GC1Q-R; GC1qR; P32; Hyaluronan Binding Protein 1; Splicing Factor SF2-Associated Protein; ASF/SF2-associated p32. Accession Number: Q07021. Gene ID: 708. Expression Region: Thr76~Gln282. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.