Learn More
Biomatik Corporation Recombinant Interferon Gamma Induced Protein 10kDa (IP10), 100UG
Supplier:  Biomatik Corporation RPU42150
Encompass_Preferred
Recombinant Human Interferon Gamma Induced Protein 10kDa (IP10) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interferon Gamma Induced Protein 10kDa (IP10). Target Synonyms: CXCL10; C7; IFI10; INP10; SCYB10; Crg2; GIP10; Mob1; Chemokine C-X-C Motif Ligand 10; Small Inducible Cytokine Subfamily B(Cys-X-Cys)Member 10. Accession Number: P02778. Gene ID: 3627. Expression Region: Val22~Pro98. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.